Ovalbumin (OVA) (Human) ELISA Kit

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB289830-100
ABCAAB289830-100EA 0 DKK
ABCAAB289830-100
Ovalbumin (OVA) (Human) ELISA Kit
Assays ELISA'er

A Sandwich ELISA kit for quantify Ovalbumin in human culture supernatants and other biological fluids.


  • Easy, convenient and time-saving method to measure the level of Ovalbumin in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids
  • Detection Range: 0,781 to 50 ng/ml
  • Sensitivity: <0,469 ng/ml
  • Assay Precision: Intra-Assay: CV <8%; Inter-Assay: CV <10%
  • This assay has high sensitivity and excellent specificity for detection of OVA. No significant cross-reactivity or interference between OVA and analogues was observed


Ovalbumin (OVA) is the main protein found in egg white, making up approximately 55% of the total protein. Ovalbumin displays sequence and three-dimensional homology to the serpin superfamily, but unlike most serpins it is not a serine protease inhibitor. The function of ovalbumin is unknown, although it is presumed to be a storage protein. It is commonly used to stimulate an allergic reaction in test subjects as well as an established model allergen for airway hyper-responsiveness (AHR). BioVision’s Ovalbumin ELISA kit is a sandwich ELISA assay for the quantitative measurement of human Ovalbumin in serum, plasma and cell culture supernatants. The density of colour is proportional to the amount of human Ovalbumin captured from the samples.

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR