DBCO-Cy3 fluorescent probe

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-[3-[1-[6-[[3-(11,12-Didehydrodibenz[b,f]azocin-5(6H)-yl)-3-oxopropyl]amino]-6-oxohexyl]-1,3-dihydro-3,3-dimethyl-5-sulpho-2H-indol-2-ylidene]-1-propen-1-yl]-3,3-dimethyl-5-sulpho-1-(3-sulphopropyl)-3H-indolium inner salt

920
AATB920EA 1140 DKK
AATB920
DBCO-Cy3 fluorescent probe
DBCO-Cy3

This azadibenzocyclooctyne-cyanine dye derivative is a versatile labeling reagent for detection of azide containing molecules or compounds.


  • Copper-free reaction
  • Azide reactive

Molekylvægt: 1185.56 g/mol
MDL Number: MFCD28505574
CAS-nummer: 1782950-79-1
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db5f3c2fb3848d86b8dd7110653952e72594eb34&vid=0d774a94508546f2a491d4ea117cf019b9a344a4&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR