ReadiCleave™ AML Cleavage Buffer, 5X Aqueous Solution, AAT Bioquest
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7510
AATB7510EA 970 DKK
AATB7510
ReadiCleave™ AML Cleavage Buffer, 5X Aqueous Solution, AAT Bioquest
Buffere Biologiske buffere
ReadiCleave™ AML Cleavage Buffer provides an optimised formula to effectively remove the fluorescence tags from ReadiCleave™ AML-labeled biological targets when needed.
  • Designed for ease of use
  • Saves time
The removal of the fluorescent ReadiCleave™ AML tags may make the fluorescence imaging more compatible with the downstream analysis of a biological target by other techniques, e.g., MS analysis.
Advarsel: For research use only.