XFD750 alkyne, FastClick™
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
72885
AATB72885EA 1990 DKK
AATB72885
XFD750 alkyne, FastClick™
XFD750 alkyne
FastClick™ XFD750 Alkyne contains both the CAG moiety of FastClick (for assisting click efficiency) and Alexa Fluor® 750 fluorophore (as the fluorescence tag) for developing Alexa Fluor® 750-based fluorescent probes.
  • Stable for 24 months
Quality tested in-house
Advarsel: For Research Use Only