Cy7 azide, FastClick™
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
72704
AATB72704EA 1990 DKK
AATB72704
Cy7 azide, FastClick™
Cy7 azide
FastClick™ Cy7 Azide contains both the CAG moiety of FastClick (for assisting click efficiency) and Cy7 fluorophore (as the fluorescence tag) for developing Cy7-based fluorescent probes.
  • Stable for 24 months
Quality tested in-house
Advarsel: For Research Use Only