MP650 Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
67019
AATB67019EA 993 DKK
AATB67019
MP650 Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Mikroplader PCR plader
MP650 is designed to have identical spectra to those of Mustang purple.
  • The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
  • This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
Advarsel: Avoid freeze-thawing plates. It is not recommended to use the plates more than 3 times. Plates can be stored for upMore Product Information