Calcein Red™ sodium salt
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
21901
AATB21901EA 2300 DKK
AATB21901
Calcein Red™ sodium salt
Calcein Red™ sodium salt
Calcein AM is one of the most popular fluorescent probes used for labeling and monitoring cellular functions of live cells.
  • In-house quality control
  • long-wavelength emission signal
  • Multiple applications
Advarsel: For research use only.