Calbryte™ 590 AM

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

20700 20701 20702
AATB20700EA 2300 DKK
AATB20700 AATB20701 AATB20702
Calbryte™ 590 AM
Calbryte™ 590 AM

The intracellular calcium flux assay is a widely used method in monitoring signal transduction pathways and high throughput screening of G protein coupled receptors (GPCRs) and calcium channel targets.


  • Improved background/signal ratio
  • Excellent intracellular retention
  • Long-wavelength emission
  • Cell permeant


Advarsel: For research use only.

Molekylvægt: 1218.77 g/mol
ADR: 5 II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR