ProLite™ His-Tag Protein Gel Staining Kit
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
18010
AATB18010EA 4240 DKK
AATB18010
ProLite™ His-Tag Protein Gel Staining Kit
Proteinekspressions Protein Ekspressionsreagenser
This kit requires very little hands-on time, allowing for rapid protein expression screening in a variety of gel types using an imaging equipment with a standard FITC filter.
  • Green fluorescence
  • Compatible with standard FITC filter
Kit provides a fast, sensitive, and highly specific fluorescent stain for visualizing His-tagged fusion proteins directly in a polyacrylamide gel following electrophoresis
It detects nanogram of His-tagged proteins directly in gels, thus eliminating the additional western blotting step.
Advarsel: For research use only.