Nuclear Orange™ DCS1
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17551
AATB17551EA 1140 DKK
AATB17551
Nuclear Orange™ DCS1
Assays Cellulær Assays
Our Nuclear Orange™ DCS1 is a fluorogenic, DNA-selective and cell-impermeant dye for analyzing DNA content in dead, fixed or apoptotic cells.
  • Cell-Impermeable
  • Useful For Multicolor Imaging
  • Bright Fluorescence
5 mM DMSO Solution
Advarsel: For research use only.