iFluor® 647 PEG12-dUTP *1 mM in TE Buffer (pH 7,5)*
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17043
AATB17043EA 1030 DKK
AATB17043
iFluor® 647 PEG12-dUTP *1 mM in TE Buffer (pH 7,5)*
Nukleinsyrereagenser Nukleotider
This iFluor® 647-PEG12-dUTP conjugate has the same spectral properties as ChromaTide™ Alexa Fluor™ 647-dUTP.
  • PEG12 spacer makes it much more effectively incorporated into DNAs by polymerase
  • Bright signal that has high photostability and is not affected by pH
The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labelled DNA via conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labelling or PCR.
Advarsel: For research use only.