iFluor® 800, Acid
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
1375
AATB1375EA 2300 DKK
AATB1375
iFluor® 800, Acid
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5 II