ReadiLink™ Rapid iFluor® 647 Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1235
AATB1235EA 1720 DKK
AATB1235
ReadiLink™ Rapid iFluor® 647 Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Proteinkoblings- og mærkningssystemer
AAT Bioquest's iFluor® dyes are optimised for labelling proteins, in particular, antibodies.
  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labelling protocol
  • No column purification required
Advarsel: For research use only.