Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (PE (Phycoerythrin)/iFluor® 594)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

116101Y0 116101Y1 116101Y2
AATB116101Y0EA 2380 DKK
AATB116101Y0 AATB116101Y1 AATB116101Y2
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (PE (Phycoerythrin)/iFluor® 594)
Antistoffer
HP-3G10 is an anti-human monoclonal antibody that recognizes the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Advarsel: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: PE (Phycoerythrin)/iFluor® 594
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR