Anti-CD38 Mouse Monoclonal Antibody [clone: HB7] (mFluor™ Violet 510)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

10382110
AATB10382110EA 2770 DKK
AATB10382110
Anti-CD38 Mouse Monoclonal Antibody [clone: HB7] (mFluor™ Violet 510)
Antistoffer
HB7 is an anti-human monoclonal antibody that recognises the CD38 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Advarsel: For research use only.

Type: Primary
Antigen: CD38 antigen
Clonality: Monoclonal
Clone: HB7
Conjugation: mFluor™ Violet 510
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR