Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 450)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

10033040 10033041
AATB10033040EA 2770 DKK
AATB10033040 AATB10033041
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 450)
Antistoffer
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Advarsel: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: iFluor® 450
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR