Alizarin red S (sodium salt), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: C.I. 58005

MQ200

A5533-25G
A5533-25GEA 1060 DKK
A5533-25G
Alizarin red S (sodium salt), Sigma-Aldrich®
Alizarin red S (sodium salt)
Formel: C₁₄H₇NaO₇S
Molekylvægt: 342.26 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013049
CAS-nummer: 130-22-3
EINECS: 204-981-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59bf7d46c0c9c3ff64b14adf84cfbe2378167bd0&vid=2168a1e5ef8eacc67bfa377aac21bdcc6957be9f&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR