X-Gal

Leverandør: PanReac AppliChem
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

A4978.0001
A4978.0001EA 2810 DKK
A4978.0001
X-Gal
X-Gal
Formel: C₁₄H₁₅BrClNO₆
Molekylvægt: 408.63 g/mol
Smeltepunkt: 237…239 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00005666
CAS-nummer: 7240-90-6
EINECS: 230-640-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR