Geranyl acetate ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (E)-3,7-Dimethylocta-2,6-dien-1-yl acetate

A19864.14 A19864.22
A19864.14EA 605 DKK
A19864.14 A19864.22
Geranyl acetate ≥98%
Geranyl acetate

Purity: 98% CAS No.: 105-87-3 Molecular Formula: (CH3)2C=CHCH2CH2C(CH3)=CHCH2O2CCH3 Formula Weight: 196.29 Storage Temperature: 2°C to 8°C Flash Point: 104°(219°F) Physical Form: Liquid Density: 0.91 Refractive Index: 1.461 Appearance: Colourless Odour: Pleasant Boiling Point: 137-139°/25mm MDL No.: MFCD00015037

Formel: C₁₂H₂₀O₂
Kogepunkt: 137…139 °C (25 mmHg)
Massefylde: 0,91
Flammepunkt: 104 °C (219 °F)
MDL Number: MFCD00015037
CAS-nummer: 105-87-3
EINECS: 203-341-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR