Phenanthren tech. 90%, Technical Grade

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A19646.0E A19646.22 A19646.36
A19646.0EEA 9780 DKK
A19646.0E A19646.22 A19646.36
Phenanthren tech. 90%, Technical Grade
Phenanthrene

Purity: 90% CAS No.: 85-01-8 Molecular Formula: C14H10 Formula Weight: 178.23 Flash Point: 171°(339°F) Melting Point: 97-101° Boiling Point: 340° Solubility: Soluble in alcohol, benzene, toluene, and glacial acetic acid Beilstein Reference: 1905428 Merck Reference: 14,7212 UN No.: 3077 MDL No.: MFCD00001168

Formel: C₁₄H₁₀
Molekylvægt: 178.23 g/mol
Kogepunkt: 340 °C
Smeltepunkt: 97…101 °C
Massefylde: 0.980
Flammepunkt: 171 °C (339 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001168
CAS-nummer: 85-01-8
EINECS: 201-581-5
UN nummer: 3077
ADR: 9 III
Merck Index: 14,7212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92527d856e1fe8582e54bf3812e1580ee0b91b05&vid=d8ec397dac001e49a7958f12aacb577cd11954da&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR