Natriumnitrit ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Diazotizing salts

A18668.0I A18668.36 A18668.0B
A18668.0IEA 1860 DKK
A18668.0I A18668.36 A18668.0B
Natriumnitrit ≥98%
Sodium nitrite

Purity: 98% CAS No.: 7632-00-0 Molecular Formula: NNaO2 Formula Weight: 69.00 Odour: Odourless Melting Point: 271° dec. UN No.: 1500 MDL No.: MFCD00011118

Formel: NaNO₂
Molekylvægt: 69 g/mol
Kogepunkt: 320
Smeltepunkt: 271 °C (dec.)
Massefylde: 2.17
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011118
CAS-nummer: 7632-00-0
EINECS: 231-555-9
UN nummer: 1500
ADR: 5.1 III
Merck Index: 13,08721

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR