2-Mercaptobenzimidazol ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Benzimidazole-2-thiol, 1,3-Dihydro-2H-benzo[d]imidazole-2-thione, Rabeprazole Related Compound C, 1,3-Dihydro-2H-benzimidazole-2-thione, 2-Benzimidazolethiol

A18350.0E A18350.22 A18350.30 A18350.36
A18350.0EEA 3170 DKK
A18350.0E A18350.22 A18350.30 A18350.36
2-Mercaptobenzimidazol ≥97%
2-Mercaptobenzimidazole

Purity: 97% CAS No.: 583-39-1 Molecular Formula: C7H6N2S Formula Weight: 150.20 Flash Point: 250°(487°F) Melting Point: 305-310° Beilstein Reference: 119867 Merck Reference: 14,1082 UN No.: 2811 MDL No.: MFCD00466107

Formel: C₇H₆N₂S
Molekylvægt: 150.2 g/mol
Kogepunkt: 376
Smeltepunkt: 305…310 °C
Massefylde: 1.42
Flammepunkt: >250 °C (487 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00466107
CAS-nummer: 583-39-1
EINECS: 209-502-6
UN nummer: 2811
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR