L(-)-Glutathion (reduceret form) ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced

A18014.06 A18014.14 A18014.22
A18014.06EA 611 DKK
A18014.06 A18014.14 A18014.22
L(-)-Glutathion (reduceret form) ≥97%
L(-)-Glutathion (reduceret form)

Purity: 97% CAS No.: 70-18-8 Molecular Formula: C10H17N3O6S Formula Weight: 307.33 Storage Temperature: 2°C to 8°C Melting Point: ca 193° dec. Beilstein Reference: 1729812 Merck Reference: 14,4475 Optical Rotation: -17° (c=2 in water) MDL No.: MFCD00065939

Formel: C₁₀H₁₇N₃O₆S
Smeltepunkt: ∼ 193 °C (dec.)
Massefylde: 1,475 g/cm³ (20 °C)
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00065939
CAS-nummer: 70-18-8
EINECS: 200-725-4
Merck Index: 14,4475

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR