2-Phenylcyclohexanon ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A17340.03 A17340.06 A17340.14
A17340.03EA 498 DKK
A17340.03 A17340.06 A17340.14
2-Phenylcyclohexanon ≥98%
2-Phenylcyclohexanon

Purity: 98% CAS No.: 1444-65-1 Molecular Formula: C12H14O Formula Weight: 174.24 Melting Point: 56-59° MDL No.: MFCD00001629

Formel: C₁₂H₁₄O
Molekylvægt: 174.24 g/mol
Kogepunkt: 90 °C (0.5 mmHg)
Smeltepunkt: 56…59 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001629
CAS-nummer: 1444-65-1
EINECS: 215-888-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR