9,10-Diethoxyanthracen ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A16705.06 A16705.03 A16705.14
A16705.06EA 1050 DKK
A16705.06 A16705.03 A16705.14
9,10-Diethoxyanthracen ≥98%
9,10-Diethoxyanthracene

Purity: 98% CAS No.: 68818-86-0 Molecular Formula: C18H18O2 Formula Weight: 266.33 Melting Point: 149-151° MDL No.: MFCD00209643

Formel: C₁₈H₁₈O₂
Smeltepunkt: 149…151 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00209643
CAS-nummer: 68818-86-0
EINECS: 272-401-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR