(-)-Dibenzoyl-L-tartaric acid, anhydrous 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (2R,3R)-2,3-Bis(benzoyloxy)succinic acid

A16181.14 A16181.22 A16181.36
A16181.14EA 358 DKK
A16181.14 A16181.22 A16181.36
(-)-Dibenzoyl-L-tartaric acid, anhydrous 99%
(-)-Dibenzoyl-L-tartaric acid

Purity: 99% CAS No.: 2743-38-6 Molecular Formula: C18H14O8 Formula Weight: 358.31 Melting Point: 154-156° MDL No.: MFCD00003074

Formel: C₁₈H₁₄O₈
Molekylvægt: 358.31 g/mol
Smeltepunkt: 154…156 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003074
CAS-nummer: 2743-38-6
EINECS: 220-374-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR