TRITON™ X-100

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

A16046.AE A16046.AP A16046.0F
A16046.AEEA 359 DKK
A16046.AE A16046.AP A16046.0F
TRITON™ X-100
TRITON™ X-100

CAS No.: 9002-93-1 Formula Weight: ca. 625 Flash Point: 180°(356°F) Physical Form: Liquid Density: 1.061 Refractive Index: 1.4900 Odour: Mild Melting Point: ca 5° Boiling Point: 250° Merck Reference: 14,6761 MDL No.: MFCD00132505


Detergent, equivalent to Nonidet P-40

Kogepunkt: >250 °C
Smeltepunkt: ∼ 5 °C
Massefylde: 1,061
Flammepunkt: 180 °C (356 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00132505
CAS-nummer: 9002-93-1
UN nummer: 3082
ADR: 9,III
Merck Index: 14,6761

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR