Benzyl-4-chlorophenyl ketone ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Benzyl 4-chlorophenyl ketone, 4'-Chloro-2-phenylacetophenone

A15959.06 A15959.14 A15959.22
A15959.06EA 583 DKK
A15959.06 A15959.14 A15959.22
Benzyl-4-chlorophenyl ketone ≥98%
Benzyl-4-chlorophenyl ketone

Purity: 98% CAS No.: 1889-71-0 Molecular Formula: C14H11ClO Formula Weight: 230.69 Melting Point: 103-107° Beilstein Reference: 976443

Formel: C₁₄H₁₁ClO
Smeltepunkt: 103…107 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00016342
CAS-nummer: 1889-71-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR