α,α,α-Trifluor-m-toluidin ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-(Trifluoromethyl)aniline, 3-Aminobenzotrifluoride

A15910.0E A15910.22 A15910.36
A15910.0EEA 3900 DKK
A15910.0E A15910.22 A15910.36
α,α,α-Trifluor-m-toluidin ≥99%
α,α,α-Trifluor-m-toluidin

Purity: 99% CAS No.: 98-16-8 Molecular Formula: C7H6F3N Formula Weight: 161.13 Flash Point: 85°(185°F) Refractive Index: 1.4800 Odour: Recognizable Melting Point: 5-6° Boiling Point: 187-189° Beilstein Reference: 387672 UN No.: 2948 MDL No.: MFCD00007797

Formel: C₇H₆F₃N
Kogepunkt: 187…189 °C
Smeltepunkt: 5…6 °C
Massefylde: 1,295
Flammepunkt: 85 °C (185 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007797
CAS-nummer: 98-16-8
EINECS: 202-643-4
UN nummer: 2948
ADR: 6.1,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR