Creatine phosphate disodium salt tetrahydrate ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Sodium creatine phosphate dibasic tetrahydrate, Sodium N-carbamimidoyl-N-methylglycinate dihydrogen phosphate tetrahydrate, Phosphocreatine disodium salt tetrahydrate

A15362.03 A15362.06 A15362.14
A15362.03EA 365 DKK
A15362.03 A15362.06 A15362.14
Creatine phosphate disodium salt tetrahydrate ≥98%
Creatine phosphate disodium salt tetrahydrate

Purity: 98+% CAS No.: 71519-72-7 Molecular Formula: C4H8N3Na2O5P.4H2O Formula Weight: 327.14 (255.07anhy) Storage Temperature: 2°C to 8°C


Substrate for creatine kinase determinations

Formel: Na₂O₃PNHC(=NH)N(CH₃)CH₂COOH·4H₂O
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00150192
CAS-nummer: 71519-72-7
EINECS: 213-074-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR