Jern (II) sulfat heptahydrat ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Iron vitriol, Ferrous sulphate heptahydrate

A15178.0C A15178.0E A15178.36
A15178.0CEA 3840 DKK
A15178.0C A15178.0E A15178.36
Jern (II) sulfat heptahydrat ≥98%
Jern (II) sulfat heptahydrat

Purity: 98% CAS No.: 7782-63-0 Molecular Formula: FeO4S.7H2O Formula Weight: 278.02 (151.91anhy) Melting Point: 64° -6H2O Merck Reference: 14,4057 MDL No.: MFCD00149719

Formel: FeSO₄·7H₂O
Molekylvægt: 278.02 g/mol
Smeltepunkt: 64 °C (-6H₂O)
Massefylde: 1.897
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149719
CAS-nummer: 7782-63-0
EINECS: 231-753-5
Merck Index: 13,04091

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR