2-(4-Hydroxyphenyl)ethanol ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol

A14891.03 A14891.06 A14891.14
A14891.03EA 403 DKK
A14891.03 A14891.06 A14891.14
2-(4-Hydroxyphenyl)ethanol ≥98%
2-(4-Hydroxyphenyl)ethanol

Purity: 98% CAS No.: 501-94-0 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 89-92° Boiling Point: 139°/1mm MDL No.: MFCD00002902

Formel: C₈H₁₀O₂
Kogepunkt: 139 °C (1 mmHg)
Smeltepunkt: 89…92 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002902
CAS-nummer: 501-94-0
EINECS: 207-930-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR