4-Brom-2-methyl-6-nitroanilin ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Bromo-6-nitro-o-toluidine, 2-Amino-5-bromo-3-nitrotoluene

A14319.14 A14319.06
A14319.14EA 2910 DKK
A14319.14 A14319.06
4-Brom-2-methyl-6-nitroanilin ≥97%
4-Brom-2-methyl-6-nitroanilin

Purity: 97% CAS No.: 77811-44-0 Molecular Formula: C7H7BrN2O2 Formula Weight: 231.06 Melting Point: 143-147° Beilstein Reference: 3530524

Formel: C₇H₇BrN₂O₂
Smeltepunkt: 143…147 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00052919
CAS-nummer: 77811-44-0
UN nummer: 3288
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR