3-Fluoranilin ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A14228.14 A14228.22 A14228.36
A14228.14EA 751 DKK
A14228.14 A14228.22 A14228.36
3-Fluoranilin ≥98%
3-Fluoranilin

Purity: 98+% CAS No.: 372-19-0 Molecular Formula: C6H6FN Formula Weight: 111.12 Flash Point: 77°(170°F) Refractive Index: 1.5440 Odour: Recognizable Boiling Point: 186-187° Beilstein Reference: 1305471 UN No.: 2941 MDL No.: MFCD00007758

Formel: C₆H₆FN
Kogepunkt: 186…187 °C
Smeltepunkt: –2 °C
Massefylde: 1,156
Flammepunkt: 77 °C (170 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007758
CAS-nummer: 372-19-0
EINECS: 206-747-0
UN nummer: 2941
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR