1,4-Dioxan ≥99% stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: p-Dioxan

Stabilization: 5-10 ppm Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

A14188.0E A14188.36
A14188.0EEA 1530 DKK
A14188.0E A14188.36
1,4-Dioxan ≥99% stabilised
1,4-Dioxan

Stabilized with ca 5-10 ppm BHT


Purity: 99+% CAS No.: 123-91-1 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: 12°(54°F) Refractive Index: 1.4220 Odour: Ether-like Melting Point: 11.8° Boiling Point: 100-102° Beilstein Reference: 102551 Merck Reference: 14,3300 UN No.: 1165 MDL No.: MFCD00006571

Formel: C₄H₈O₂
Molekylvægt: 88.11 g/mol
Kogepunkt: 100…102 °C
Smeltepunkt: 11.8
Massefylde: 1.034
Flammepunkt: 12 °C (54 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006571
CAS-nummer: 123-91-1
EINECS: 204-661-8
UN nummer: 1165
ADR: 3 II
Merck Index: 14,3300
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR