4-Brom-3,5-dimethylisoxazol ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A14179.03 A14179.06 A14179.14
A14179.03EA 498 DKK
A14179.03 A14179.06 A14179.14
4-Brom-3,5-dimethylisoxazol ≥97%
4-Bromo-3,5-dimethylisoxazole

Purity: 97% CAS No.: 10558-25-5 Molecular Formula: C5H6BrNO Formula Weight: 176.02 Storage Temperature: 2°C to 8°C Flash Point: 73°(163°F) Refractive Index: 1.4880 Boiling Point: 176° Beilstein Reference: 108522 MDL No.: MFCD00068187

Formel: C₅H₆BrNO
Molekylvægt: 176.01 g/mol
Kogepunkt: 176 °C
Massefylde: 1.49
Flammepunkt: 73 °C (163 °F)
MDL Number: MFCD00068187
CAS-nummer: 10558-25-5

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR