1,2-Dichlor-4-iodbenzen ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3,4-Dichloroiodobenzene

A13440.09 A13440.18
A13440.09EA 451 DKK
A13440.09 A13440.18
1,2-Dichlor-4-iodbenzen ≥98%
1,2-Dichlor-4-iodbenzen

Purity: 98% CAS No.: 20555-91-3 Molecular Formula: C6H3Cl2I Formula Weight: 272.90 Refractive Index: 1.6480 Melting Point: 29-31° Boiling Point: 116-118°/12mm Beilstein Reference: 2245136 MDL No.: MFCD00019014

Formel: C₆H₃Cl₂I
Kogepunkt: 116…118 °C (12 mmHg)
Smeltepunkt: 29…31 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00019014
CAS-nummer: 20555-91-3
EINECS: 243-874-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR