2-Brom-1,1-diethoxyethan ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Bromo-1,1-diethoxyethane

A13276.0E A13276.22 A13276.36
A13276.0EEA 4890 DKK
A13276.0E A13276.22 A13276.36
2-Brom-1,1-diethoxyethan ≥97%
2-Brom-1,1-diethoxyethan

Purity: 97% CAS No.: 2032-35-1 Molecular Formula: C6H13BrO2 Formula Weight: 197.08 Storage Temperature: 2°C to 8°C Flash Point: 61°(142°F) Refractive Index: 1.4390 Boiling Point: 66-67°/18mm Beilstein Reference: 635754 UN No.: 2810 MDL No.: MFCD00000214

Formel: BrH₂CCH(OCH₂CH₃)₂
Kogepunkt: 66…67 °C (18 mmHg)
Massefylde: 1,270
Flammepunkt: 61 °C (142 °F)
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00000214
CAS-nummer: 2032-35-1
EINECS: 217-989-1
UN nummer: 2810
ADR: 6.1,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR