9,10-Anthraquinon 98

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Anthracene-9,10-dione, Anthraquinone

A12735.30
A12735.30EA 1060 DKK
A12735.30
9,10-Anthraquinon 98
9,10-Anthraquinon

Purity: 98+% CAS No.: 84-65-1 Molecular Formula: C14H8O2 Formula Weight: 208.22 Flash Point: 185°(365°F) Odour: Odourless Boiling Point: 377° Beilstein Reference: 390030 Merck Reference: 14,687 MDL No.: MFCD00001188

Formel: C₁₄H₈O₂
Molekylvægt: 208.22 g/mol
Kogepunkt: 377 °C
Smeltepunkt: 283…287 °C
Massefylde: 1.438
Flammepunkt: 185 °C (365 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001188
CAS-nummer: 84-65-1
EINECS: 201-549-0
Merck Index: 13,00692

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR