Kaliumiodid ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Dipotassium diiodide, Kali iodide

A12704.0B A12704.0I A12704.18 A12704.30
A12704.0BEA 3160 DKK
A12704.0B A12704.0I A12704.18 A12704.30
Kaliumiodid ≥99%
Potassium iodide

Purity: 99% CAS No.: 7681-11-0 Molecular Formula: KI Formula Weight: 166.01 Refractive Index: 1.677 Odour: Odourless Boiling Point: 1330° Merck Reference: 14,7643 MDL No.: MFCD00011405

Formel: Kl
Molekylvægt: 166 g/mol
Kogepunkt: 1330 °C
Smeltepunkt: 681 °C
Massefylde: 3.12
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011405
CAS-nummer: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR