2,2'-Bithiophen ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2,2′-Dithienyl, 2,2′-Bithienyl

A12335.03 A12335.06 A12335.14
A12335.03EA 465 DKK
A12335.03 A12335.06 A12335.14
2,2'-Bithiophen ≥98%
2,2'-Bithiophen

Purity: 98% CAS No.: 492-97-7 Molecular Formula: C8H6S2 Formula Weight: 166.27 Flash Point: 110°(230°F) Melting Point: 30-33° Boiling Point: 260° Beilstein Reference: 3039 MDL No.: MFCD00005414

Formel: C₈H₆S₂
Kogepunkt: 260 °C
Smeltepunkt: 30…33 °C
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005414
CAS-nummer: 492-97-7
EINECS: 207-767-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR