Natriummetaborat tetrahydrat ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A11803.0E A11803.0C A11803.36
A11803.0EEA 1700 DKK
A11803.0E A11803.0C A11803.36
Natriummetaborat tetrahydrat ≥98%
Natriummetaborat tetrahydrat

Purity: 98% CAS No.: 10555-76-7 Molecular Formula: NaBO2.4H2O Formula Weight: 137.86 (65.80anhy) Odour: Odourless Melting Point: ca 53° dec. MDL No.: MFCD00149205

Formel: BNaO₂·4H₂O
Molekylvægt: 137.86 g/mol
Smeltepunkt: ∼ 53 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149205
CAS-nummer: 10555-76-7
EINECS: 231-891-6

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR