4,4'-Dimethoxybenzophenon ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Bis(4-methoxyphenyl)methanone

A11466.14 A11466.22 A11466.36
A11466.14EA 627 DKK
A11466.14 A11466.22 A11466.36
4,4'-Dimethoxybenzophenon ≥98%
4,4'-Dimethoxybenzophenon

Purity: 98+% CAS No.: 90-96-0 Molecular Formula: C15H14O3 Formula Weight: 242.27 Melting Point: 142-146° Beilstein Reference: 1878026

Formel: C₁₅H₁₄O₃
Kogepunkt: 180…240 °C (13 mmHg)
Smeltepunkt: 142…146 °C
Massefylde: 1,259 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008404
CAS-nummer: 90-96-0
EINECS: 202-028-0

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR