Thiamin hydrochlorid

Leverandør: PanReac AppliChem
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride

A0955.0250
A0955.0250EA 2450 DKK
A0955.0250
Thiamin hydrochlorid
Thiamin hydrochlorid
Formel: C₁₂H₁₈Cl₂N₄OS
Molekylvægt: 337.27 g/mol
Smeltepunkt: 252
Massefylde: 1.401 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012780
CAS-nummer: 67-03-8
EINECS: 200-641-8
Merck Index: 13,09366

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR