Xylen, BAKER ANALYZED®, Laboratory Reagent Spectrophotometry, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Dimethylbenzene

9516.1000EA 733 DKK
9516.1000
Xylen, BAKER ANALYZED®, Laboratory Reagent Spectrophotometry, J.T.Baker®
Xylen
Formel: C₆H₄(CH₃)₂
Molekylvægt: 106.17 g/mol
Kogepunkt: 140
Smeltepunkt: –34 °C
Massefylde: 0.86…0.88 g/cm³ (20 °C)
Flammepunkt: 25
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00077264
CAS-nummer: 1330-20-7
EINECS: 215-535-7
UN nummer: 1307
ADR: 3 III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Boiling Range (initial to dry point) 137 - 144 °C
Color (APHA) max. 10
Free Acid (µeq/g) max. 0.12
Residue after Evaporation max. 0.001 %
Sulfur Compounds (as S) max. 0.003 %
Substances Darkened by H2SO4 passes test
Water (H2O) max. 0.02 %
Ultraviolet Absorbance (1.00-cm path vs water):
at 295 nm max. 1.00
at 300 nm max. 0.30
at 330 nm max. 0.05
at 380-400 nm max. 0.01


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR