Borsyre 40 g/l in aqueous solution med indikator, VOLUSOL®

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

95097.5000EA 1080 DKK
95097.5000
Borsyre 40 g/l in aqueous solution med indikator, VOLUSOL®
Borsyre
Formel: B(OH)₃
Molekylvægt: 61.83 g/mol
Kogepunkt: 185
Smeltepunkt: 160
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011337
CAS-nummer: 10043-35-3
EINECS: 233-139-2
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay (W/V) 3.9 - 4.2 %
Appearance Clear red liquid
Sensitivity Passes test
Density (20/4) 1.000 - 1.020

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR