Natriumnitrit solution, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Diazotizing salts

914-10ML
914-10MLEA 760 DKK
914-10ML
Natriumnitrit solution, Sigma-Aldrich®
Sodium nitrite

For use in the Naphthol AS-D Chloroacetate Esterase and Naphthyl Acetate Esterase Procedures, Kits 91C and 91A.

Formel: NaNO₂
Molekylvægt: 69 g/mol
Kogepunkt: 320
Smeltepunkt: 284
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00011118
CAS-nummer: 7632-00-0
Merck Index: 13,08721
IVDLogo

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR