Kaliumpermanganat concentrate for dilution to 0.002 mol (0.01N) aqueous solution, ConvoL NORMADOSE® Volumetric

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Permanganic acid potassium salt

conc_1l

91353.600EA 445 DKK
91353.600
Kaliumpermanganat concentrate for dilution to 0.002 mol (0.01N) aqueous solution, ConvoL NORMADOSE® Volumetric
Potassium permanganate
Formel: KMnO₄
Molekylvægt: 158.03 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011364
CAS-nummer: 7722-64-7
Merck Index: 12,07824

Bestil nu

Specifikationer Testresultater

Molarity 0.00199 - 0.00201 mol/l (20°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR