5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) ≥99% (by TLC) amine-reactive fluorescent dye

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2,5-Dioxopyrrolidin-1-yl 3',6'-dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylate, 5-FAM-N-succinimidyl ester, 5-Carboxyfluorescein NHS ester, 5-FAM SE

90029
90029.EA 1730 DKK
90029.
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) ≥99% (by TLC) amine-reactive fluorescent dye
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE)

5-FAM SE is an amine-reactive form of 5-carboxyfluorescein single isomer.


  • λEx/λEm (pH 9) = 495/519 nm
  • Yellow orange solid soluble in DMF or DMSO
  • Store at -20°C and protect from light

Formel: C₂₅H₁₅NO₉
Molekylvægt: 473.39 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00077322
CAS-nummer: 92557-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e17edacd0d3e4887beccf7c05605b1d6be017e2&vid=4e17edacd0d3e4887beccf7c05605b1d6be017e2&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR