2,6-Dichlorquinon-4-chlorimid 98, AnalaR NORMAPUR®

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

88253.120EA 810 DKK
88253.120
2,6-Dichlorquinon-4-chlorimid 98, AnalaR NORMAPUR®
2,6-Dichlorquinon-4-chlorimid
Formel: C₆H₂Cl₃NO
MDL Number: MFCD00001611
CAS-nummer: 101-38-2
UN nummer: 3224
ADR: 4.1

Bestil nu

Specifikationer Testresultater

Assay ≥ 98 %
Appearance: Brownish-yellow crystalline powder Passes test
Melting point 65 - 68 °C
Spec.absorptivity (312 nm; 0.001%; ethanol) ≥ 800


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR